Key Documents
Z126055
Ace McLeod vacuum gauge
pressure 0-10 mmHg
Select a Size
1 EA
$2,050.00
$2,050.00
Estimated to ship onMarch 11, 2025
Select a Size
Change View
1 EA
$2,050.00
About This Item
UNSPSC Code:
41112406
NACRES:
NB.79
$2,050.00
Estimated to ship onMarch 11, 2025
manufacturer/tradename
Ace Glass 872807
parameter
0-10 mmHg pressure
General description
With interchangeable measuring tube. Mercury not included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)