Key Documents
Z106410
Supelco
Black plastic caps
Solid top, white rubber liner, thread for 15-425, pkg of 200 ea
Select a Size
200 EA
$124.00
$124.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
200 EA
$124.00
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
$124.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
material
black phenolic cap
white rubber liner
feature
closure type black phenolic solid top cap
packaging
pkg of 200 ea
NMR tube diam.
10 mm
fitting
thread for 15-425
General description
These autoclavable caps are designed to ensure the ultimate in product safety.
Choose from one of the most recent versions:
Certificates of Analysis (COA)