As mentioned on the Product Detail Page in the PROPERTIES section, the platform size is 20 cm × 20 cm. It has a vertical distance of 18 cm and a max working height of 26 cm.
Select a Size
$183.00
Select a Size
About This Item
$183.00
Recommended Products
feature
capacity≤ 100 lb static load
manufacturer/tradename
Grauer LBJSET
platform size
20 cm × 20 cm
support rod L
45 cm
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How big is the platform?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service