Key Documents
Z104574
Scienceware® flow indicator
gives immediate visual indication
Select a Size
1 EA
$41.90
$41.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$41.90
About This Item
UNSPSC Code:
20121904
NACRES:
NB.51
$41.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art H19935-0000
General description
Gives immediate visual indication of cooling-water (or gas) flow by rotation of white ball in the circular channel. A minimum water flow of 50 mL/min (indicator in horizontal position) or 140 mL/min (indicator in vertical position) will actuate the ball. With air, the corresponding minimum flow rates are 3 and 8 L/min. Maximum pressure is 50 psig with water and 10 psig with air.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)