Key Documents
Z104353
Sigma-Aldrich
Tubing quick-disconnects
for tubing i.d., 1/4 ‑ 5/16 in.
Select a Size
12 EA
$58.80
$58.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
12 EA
$58.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$58.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PE
feature
for tubing i.d. 1/4 ‑ 5/16 in.
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art H19728-0000
General description
Consists of two serrated connectors which are joined securely through a male-female center taper. Connector of one size can be interchanged with other sizes to produce reducing combinations.
  • Easy-to-assemble barbed connectors require no tools
  • Ideal for use with flexible tubing
  • Handy on lines that require frequent disconnection
Choose from one of the most recent versions:
Certificates of Analysis (COA)