Key Documents
Z103098
Sigma-Aldrich
Aldrich® dry ice condenser
joint: ST/NS 24/40
Select a Size
1 EA
$1,130.00
$1,130.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
Select a Size
Change View
1 EA
$1,130.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$1,130.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onMay 31, 2025
Joint
joint: ST/NS24/40
General description
For use with dry ice/acetone slush or liquid nitrogen in cold-finger.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)