Key Documents
Z101923
Sigma-Aldrich
Single-bulb ball-tube
capacity 500 mL, joint: ST/NS 24/40, plain
Select a Size
1 EA
$236.00
$236.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$236.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
$236.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
capacity
500 mL
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Choose from one of the most recent versions:
Certificates of Analysis (COA)