Key Documents
Z100307
Sigma-Aldrich
Aldrich® West condenser w/ Clear-Seal joint
joint: ST/NS 19/22, pkg of 1 ea
Select a Size
1 EA
$192.00
$192.00
Estimated to ship onMay 27, 2025
Select a Size
Change View
1 EA
$192.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$192.00
Estimated to ship onMay 27, 2025
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
L
200 mm
General description
Replacement part for the Diazald Kit, Z100250.
The Diazald kit is a set of distillation glassware designed for the safe preparation of diazomethane (~100 mmol). Glassware without sharp edges or ground-glass joints are recommended by de Boer and Backer [Org. Syn., Coll. Vol. 4, 250 (1963)]. The Diazald kit features 19/22 Clear-Seal joints; these joints do not require grease even for vacuum applications, thus avoiding a source of contamination.
The glassware in the kit should be washed with care and without the use of wire brushes which will scratch the inner surface. For further information, request Technical Information Bulletin No. AL-180 and for a review on the preparation and reactions of diazomethane, see Aldrichim. Acta, 16, 1, 1983.
Call or visit our website for pricing on replacement parts.
NOTE: We are unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)