Key Documents
WHAWB100024
Qiagen® Multi Barrier Pouch
large, size , L x W 7" x 7.37" (17.8 cm x 18.7 cm), resealable, pkg of 50 ea
Synonym(s):
Multi Barrier Pouch Reseal 7 x 7.37 (50), Whatman FTA, Z747351
About This Item
UNSPSC Code:
41105500
NACRES:
NB.22
Pricing and availability is not currently available.
packaging
pkg of 50 ea
manufacturer/tradename
Qiagen WB100024
size
, L x W 7" x 7.37" (17.8 cm x 18.7 cm)
Related Categories
General description
Large size 7 x 7.37 inch (17.8 x 18.7 cm) resealable Multi-Barrier Pouches for transport or storage of QIAcard FTA CloneSaver.
Features and Benefits
  • Laminated layers maintain sample integrity and security by protecting the card from exposure to gas or liquid contamination.
  • The outer paper surface is suitable for labeling or writing. The Multi-Barrier Pouch has a resealable closure. To optimize sample integrity FTA Cards are stored in a Multi-Barrier Pouch with a desiccant packet.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Qiagen is a registered trademark of Qiagen Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Description of common problems that can occur during genomic DNA preparation
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.
Protocols
A number of options are available for sample collection, transport, archiving, and DNA purification.
Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.