WHA68912507
Whatman® GD/X syringe filters
GF/F membrane, pore size 0.7 μm, diam. 25 mm, non-sterile, pkg of 1500 ea
Synonym(s):
Whatman filter, Z745502, syringe filter, syringe filters
About This Item
Recommended Products
material
glass microfiber membrane
polypropylene housing
sterility
non-sterile
feature
glass fiber prefilter
packaging
pkg of 1500 ea
manufacturer/tradename
Whatman 6891-2507
Whatman Article No. 28420543 (US reference)
diam.
25 mm
volume
≤25 mL
pore size
0.7 μm
fitting
female Luer inlet (lock)
male Luer outlet
Looking for similar products? Visit Product Comparison Guide
Related Categories
1 of 4
This Item | WHA68712502 | WHA68952504 | WHA68902507 |
---|---|---|---|
manufacturer/tradename Whatman 6891-2507, Whatman Article No. 28420543 (US reference) | manufacturer/tradename Whatman 6871-2502, Whatman Article No. 28420496 (US reference) | manufacturer/tradename Whatman 6895-2504, Whatman Article No. 28420547 (US reference) | manufacturer/tradename Whatman 6890-2507, Whatman Article No. 28420542 (US reference) |
pore size 0.7 μm | pore size 0.2 μm | pore size 0.45 μm | pore size 0.7 μm |
material glass microfiber membrane, polypropylene housing | material Nylon membrane, polypropylene housing | material glass microfiber membrane, polypropylene housing | material glass microfiber membrane, polypropylene housing |
fitting female Luer inlet (lock), male Luer outlet | fitting female Luer inlet (lock), male Luer outlet | fitting female Luer inlet (lock), male Luer outlet | fitting female Luer inlet (lock), male Luer outlet |
sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
General description
Features and Benefits
- Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service