WHA2810266
Whatman® high performance cellulose extraction thimbles
double thickness, L × W 26 mm × 60 mm
Synonym(s):
Z753513
About This Item
Recommended Products
packaging
pk of 25
manufacturer/tradename
Whatman 2810-266
Whatman Article No. 28419008 (US reference)
L × W
26 mm × 60 mm
Related Categories
General description
Cellulose thimble options for Soxhlet extractor systems
Standard extraction thimbles have a wall thickness of 1–1.5 mm. Double-thickness thimbles have a wall thickness of approximately 2 mm for applications that require higher retention, increased wet or dry strength, or increased rigidity. Measurements can be matched to specific Soxhlet extractor systems.
Features and Benefits
- Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
- Suitable for temperatures up to 120°C (248°F)
- Excellent mechanical strength and retention
- Wall thickness of standard thimbles is 1–1.5 mm to support application need
- Option for double thickness for added rigidity
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)