WHA1091110
Whatman® qualitative filter paper, Grade 91, creped
circles, diam. 110 mm, pack of 4000
Synonym(s):
Z752444
About This Item
Recommended Products
material
cellulose filters
circles
packaging
pack of 4000
manufacturer/tradename
Whatman 1091-110
Whatman Article No. 28417530 (US reference)
parameter
2 psi wet burst
70 sec/100 mL speed (Herzberg)
diam.
110 mm
thickness
205 μm
pore size
10 μm (Particle retention)
basis weight
71 g/m2
Looking for similar products? Visit Product Comparison Guide
General description
Grade 91: 10 μm (credped filter paper)
A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration.
Features and Benefits
- A general purpose creped filter for less critical routine analysis.
- Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service