Skip to Content
Merck
All Photos(1)

Key Documents

WHA10538018

Whatman® qualitative filter paper, Grade CF 12

sheets, W × L 580 mm × 580 mm, pack of 100

Synonym(s):

filter paper, whatman filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104929
NACRES:
NB.24
Pricing and availability is not currently available.

material

sheets

feature

Grade CF 12

packaging

pack of 100

manufacturer/tradename

Whatman 10538018
Whatman Article No. 28417051 (US reference)

W × L

580 mm × 580 mm

Looking for similar products? Visit Product Comparison Guide

General description

This high-quality filter paper is ideal for applications requiring efficient filtration and separation of fine particles, ensuring reliable results in laboratory settings.

Features and Benefits

  • Grade CF 12
  • 580 mm × 580 mm

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service