WHA10423029
Whatman® TG100 filter discs 80 μm pore size
100 ea, 50 mm diam
About This Item
Recommended Products
Product Name
Whatman® Separation Gauze, diam. 50 mm, pore size 80 μm, pack of 100
packaging
pack of 100
manufacturer/tradename
Whatman 10423029
diam.
50 mm
pore size
80 μm pore size
Related Categories
Features and Benefits
- Minimal sample loss in protein-based samples due to low protein binding
- Pure cellulose acetate composition, minimizing sample contamination
- Hydrophilic, for use with aqueous and alcoholic media
- High solvent resistance, particularly to low molecular weight alcohols
- High physical strength and a heat resistance up to 180°C
- Can be sterilized by most common methods without sacrificing membrane integrity
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service