Skip to Content
Merck
All Photos(3)

Key Documents

W800075

Sigma-Aldrich

Linoleic acid

greener alternative

natural (US), ≥95%, FG

Synonym(s):

cis-9,cis-12-Octadecadienoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

W800075-SAMPLE
$62.20
100 G
$141.00
1 KG
$821.00

$62.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W800075-SAMPLE
$62.20
100 G
$141.00
1 KG
$821.00

About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
280.45
Beilstein:
1727101
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
natural (US)
biological source:
(Sunflower seed)
food allergen:
no known allergens

$62.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

(Sunflower seed)

Quality Level

300
400

grade

FG
Fragrance grade
Halal
natural (US)

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117
FDA 21 CFR 184.1065

Assay

≥95%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.466 (lit.)

bp

229-230 °C/16 mmHg (lit.)

mp

−5 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

fatty

SMILES string

OC(CCCCCCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-

InChI key

OYHQOLUKZRVURQ-HZJYTTRNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Biochem/physiol Actions

Linoleic acid increases cell proliferation and gene expression of PPARα and its target genes such as acyl-CoA oxidase in primary duck hepatocytes [1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is it possible to synthesize linoleic acid synthetic? What I mean is, is it possible to make linoleic acid that does not come from a natural source?

    1 answer
    1. Currently, all linoleic acid products are purified from biological sources. This is the most common source for commercial linoleic acid.
      It may be possible to synthesize linoleic acid.

      To inquire into custom manufacturing capabilities, we kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Quotes" under the Orders section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service