Skip to Content
Merck
All Photos(1)

Documents

W530183

Sigma-Aldrich

Petitgrain oil, Paraguay

Certified organic (NOP)

Synonym(s):

Bitter orange oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2855
MDL number:
UNSPSC Code:
12164502

grade

Certified organic (NOP)
Halal
Kosher

origin

Paraguay origin

refractive index

n20/D 1.459 (lit.)

bp

173 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

152.0 °F

Flash Point(C)

66.67 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service