About This Item
Recommended Products
biological source
synthetic
Assay
>96%
refractive index
n20/D 1.496 (lit.)
bp
259 °C (lit.)
density
0.976 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Organoleptic
woody
SMILES string
CO[C@]1(C)CC[C@@]23CC1C(C)(C)C2CC[C@H]3C
InChI
1S/C16H28O/c1-11-6-7-12-14(2,3)13-10-16(11,12)9-8-15(13,4)17-5/h11-13H,6-10H2,1-5H3/t11-,12+,13-,15-,16+/m1/s1
InChI key
HRGPYCVTDOECMG-RHBQXOTJSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service