Skip to Content
Merck
All Photos(1)

Key Documents

W525308

Sigma-Aldrich

Nerolin Yara Yara

≥99%

Synonym(s):

2-Methoxynaphthalene, Methyl 2-naphthyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH3
CAS Number:
Molecular Weight:
158.20
FEMA Number:
4704
Beilstein:
1859408
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.074

biological source

synthetic

Assay

≥99%

form

flakes
powder

origin

China origin

bp

274 °C (lit.)

mp

70-73 °C (lit.)

density

1.064 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; strawberry; sweet

SMILES string

COc1ccc2ccccc2c1

InChI

1S/C11H10O/c1-12-11-7-6-9-4-2-3-5-10(9)8-11/h2-8H,1H3

InChI key

LUZDYPLAQQGJEA-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Widén et al.
Food additives and contaminants, 22(7), 681-692 (2005-07-16)
Mineral water and soft drinks with a perceptible off-odour were analysed to identify contaminants originating from previous misuse of the refillable polyethylene terephthalate (PET) bottle. Consumers detected the off-odour after opening the bottle and duly returned it with the remaining
R R Price et al.
Journal of microencapsulation, 10(2), 215-222 (1993-04-01)
Many natural products that exhibit biocidal activity have poor solubility in water. In order to explore the prolonged delivery of these compounds from microtubules we have utilized 2-methoxynaphthalene as a model to elucidate release characteristics of hydrophobic compounds entrapped in
Mingzhang Gao et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 68(3), 459-465 (2010-01-12)
Carbon-11-labeled piperidine ring of N-[omega-(6-methoxynaphthalen-1-yl)alkyl] derivatives were first designed and synthesized as new selective PET sigma(1) receptor probes. The target tracers were prepared by O-[(11)C]methylation of their corresponding phenolic hydroxyl precursors using [(11)C]CH(3)OTf under basic conditions and isolated by a
G M Whited et al.
Bioorganic & medicinal chemistry, 2(7), 727-734 (1994-07-01)
2-Methoxynaphthalene was subjected to biooxidation by whole cells of six organisms: Pseudomonas putida F39/D containing toluene dioxygenase, Escherichia coli JM109(pDTG601), containing recombinant toluene dioxygenase from Pp F39/D, Pseudomonas sp. NCIB 9816/11, containing naphthalene dioxygenase. E. coli JM109(pDTG141), containing recombinant naphthalene
P Di Gennaro et al.
Biotechnology and bioengineering, 93(3), 511-518 (2005-09-30)
The bioconversion of naphthalene to the 1,2-dihydro-1,2-dihydroxy derivative was performed in good yield using an Escherichia coli recombinant strain carrying Pseudomonas fluorescens N3 dioxygenase. However, the efficiency of such transformation is affected by many process parameters, and their optimization is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service