Skip to Content
Merck
All Photos(1)

Key Documents

W520608

Sigma-Aldrich

1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran solution

in diethyl phthalate

Synonym(s):

HHCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O
CAS Number:
Molecular Weight:
258.40
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
floral; sweet
grade:
Fragrance grade
biological source:
synthetic
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

concentration

48-52% in diethyl phthalate

refractive index

n20/D 1.5215 (lit.)

bp

304 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

floral; sweet

SMILES string

CC(C(C)C1(C)C)(C)C2=C1C=C(C(C)OCC3)C3=C2

InChI

1S/C18H26O/c1-11-14-10-16-15(9-13(14)7-8-19-11)17(3,4)12(2)18(16,5)6/h9-12H,7-8H2,1-6H3

InChI key

YWMTVUUYOAIDBC-UHFFFAOYSA-N

General description

1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran is an isochromone with a strong musk odor,[1] commonly used in fragrances.[2]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and olfactory characterization of the enantiomerically pure isomers of the perfumery synthetic Galaxolide?.
Frater G, et al.
Helvetica Chimica Acta, 82(10), 1656-1665 (1999)
Ursula Klaschka et al.
Environmental science and pollution research international, 20(4), 2456-2471 (2012-09-05)
Fragrances are used in a wide array of everyday products and enter the aquatic environment via wastewater. While several musk compounds have been studied in detail, little is known about the occurrence and fate of other fragrances. We selected 16
Shuo Liu et al.
Chemosphere, 83(8), 1080-1086 (2011-02-22)
Although polycyclic musks have been shown to cause lethal and sub-lethal effects on organisms, their biochemical toxicity to earthworms is not well understood. In the current study, we investigated the responses of antioxidant systems and lipid peroxidation after exposing Eisenia
Cui-Hong Chen et al.
Huan jing ke xue= Huanjing kexue, 32(2), 567-573 (2011-05-03)
Using outdoor pot-culture experiment,biomasses of wheat seedlings and accumulation of HHCB and/or Cd in parts of wheat seedlings cultured in alluvial soil and cinnamon soil were investigated. The biomasses of wheat seedlings in different treatments followed the order as single
Evaluation of the oral subchronic toxicity of HHCB (1, 3, 4, 6, 7, 8-hexahydro-4, 6, 6, 7, 8, 8-hexamethylcyclopenta-?-2-benzopyran) in the rat.
Api AM and Ford RA
Toxicology Letters, 111(1), 143-149 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service