Skip to Content
Merck
All Photos(1)

Documents

W513105

Sigma-Aldrich

m-Anisaldehyde

≥97%

Synonym(s):

3-Methoxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
Beilstein:
606013
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.158
NACRES:
NA.21

biological source

synthetic

reg. compliance

FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.553 (lit.)

bp

143 °C/50 mmHg (lit.)

density

1.117 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

anise

SMILES string

[H]C(=O)c1cccc(OC)c1

InChI

1S/C8H8O2/c1-10-8-4-2-3-7(5-8)6-9/h2-6H,1H3

InChI key

WMPDAIZRQDCGFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Anisaldehyde is a flavoring food additive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural and spectroscopic (UV-Vis, IR, Raman, and NMR) characteristics of anisaldehydes that are flavoring food additives: A density functional study in comparison with experiments.
Altun A, et al
Journal of Molecular Structure, 1128, 590-605 (2017)
Chao-Bin Xue et al.
Bioorganic & medicinal chemistry, 15(5), 2006-2015 (2007-01-30)
Phenoloxidase (PO), also known as tyrosinase, is a key enzyme in insect development, responsible for catalyzing the hydroxylation of tyrosine into o-diphenols and the oxidation of o-diphenols into o-quinones. Inhibition of PO may provide a basis for novel environmentally friendly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service