Skip to Content
Merck
All Photos(1)

Key Documents

W510424

Sigma-Aldrich

3′,4′-Dimethoxyacetophenone

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
(CH3O)2C6H3COCH3
CAS Number:
Molecular Weight:
180.20
Beilstein:
781213
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Assay

≥98%

bp

286-288 °C (lit.)

mp

47-54 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

woody; floral; sweet

SMILES string

COc1ccc(cc1OC)C(C)=O

InChI

1S/C10H12O3/c1-7(11)8-4-5-9(12-2)10(6-8)13-3/h4-6H,1-3H3

InChI key

IQZLUWLMQNGTIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service