Skip to Content
Merck
All Photos(1)

Key Documents

W501301

Sigma-Aldrich

Ethyl 2-furoate

≥98%, FG

Synonym(s):

Ethyl furan-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
Beilstein:
2653
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.122
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

bp

196 °C (lit.)

mp

32-37 °C (lit.)

density

1.117 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; balsamic; fruity

SMILES string

CCOC(=O)c1ccco1

InChI

1S/C7H8O3/c1-2-9-7(8)6-4-3-5-10-6/h3-5H,2H2,1H3

InChI key

NHXSTXWKZVAVOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 2-furoate is found in apple cider, mango wines, citrus jam, dried apricot, dried figs, prune, honey and coconut. It is used as a flavoring and fragrance agent in industries.[1][2][3] Ethyl 2-furoate can be synthesized by Fischer esterification reaction between 2-furoic acid and ethanol using acid catalysts.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomass valorization derivatives: clean esterification of 2-furoic acid using tungstophosphoric acid/zirconia composites as recyclable catalyst
Escobar A, et al.
Process Safety and Environmental Protection, 98, 176-186 (2015)
Sensory properties and aroma compounds of sweet Fiano wine
Genovese A, et al.
Food Chemistry, 103(4), 1228-1236 (2007)
Wines from fruits other than grapes: Current status and future prospectus
Jagtap UB and Bapat VA
Food Bioscience, 9, 80-96 (2015)
Analysis of volatile compounds of mango wine.
Pino JA & Queris O.
Food Chemistry, 125(4), 1141-1146 (2011)
Nadhem Aissani et al.
Journal of agricultural and food chemistry, 63(27), 6120-6125 (2015-06-18)
Research on new pesticides based on plant extracts, aimed at the development of nontoxic formulates, has recently gained increased interest. This study investigated the use of the volatilome of rucola (Eruca sativa) as a powerful natural nematicidal agent against the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service