Skip to Content
Merck
All Photos(1)

Key Documents

W444401

Sigma-Aldrich

Ethyl 5-hydroxydecanoate

solution in delta-decalactone

Sign Into View Organizational & Contract Pricing

Select a Size

W444401-SAMPLE-K
$78.10
100 G
$324.00
1 KG
$1,250.00

$78.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W444401-SAMPLE-K
$78.10
100 G
$324.00
1 KG
$1,250.00

About This Item

Empirical Formula (Hill Notation):
C12H24O3
CAS Number:
Molecular Weight:
216.32
FEMA Number:
4444
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

$78.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Kosher

concentration

≥55%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; peach; sweet

SMILES string

O(CC)C(=O)CCCC(O)CCCCC

InChI

1S/C12H24O3/c1-3-5-6-8-11(13)9-7-10-12(14)15-4-2/h11,13H,3-10H2,1-2H3

InChI key

JZHPHZPSKLWKPM-UHFFFAOYSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service