Skip to Content
Merck
All Photos(1)

Key Documents

W424101

Sigma-Aldrich

Hexylamine

99%

Synonym(s):

1-Aminohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5NH2
CAS Number:
Molecular Weight:
101.19
FEMA Number:
4241
Beilstein:
1731298
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.016

Assay

99%

expl. lim.

2.1-9.3 %

refractive index

n20/D 1.418 (lit.)

bp

131-132 °C (lit.)

mp

−23 °C (lit.)

density

0.766 g/mL at 25 °C (lit.)

SMILES string

CCCCCCN

InChI

1S/C6H15N/c1-2-3-4-5-6-7/h2-7H2,1H3

InChI key

BMVXCPBXGZKUPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service