W407701
o-Anisaldehyde
≥97%, FG
Synonym(s):
2-Methoxybenzaldehyde, o-Anisaldehyde, Salicylaldehyde methyl ether
About This Item
Recommended Products
biological source
synthetic
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515
Assay
≥97%
bp
238 °C (lit.)
mp
34-40 °C (lit.)
density
1.127 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
almond; hawthorne; sweet; vanilla
SMILES string
[H]C(=O)c1ccccc1OC
InChI
1S/C8H8O2/c1-10-8-5-3-2-4-7(8)6-9/h2-6H,1H3
InChI key
PKZJLOCLABXVMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
244.4 °F - closed cup
Flash Point(C)
118 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service