W390003
Sodium diacetate
Synonym(s):
Sodium hydrogen diacetate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H7NaO4
CAS Number:
Molecular Weight:
142.09
FEMA Number:
3900
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.073
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens
Recommended Products
biological source
synthetic
Quality Level
reg. compliance
FCC
FDA 21 CFR 184.1754
form
solid
mp
323-329 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
faint
SMILES string
[Na+].CC(O)=O.CC([O-])=O
InChI
1S/2C2H4O2.Na/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+1/p-1
InChI key
BHZOKUMUHVTPBX-UHFFFAOYSA-M
General description
Sodium diacetate is a flavoring compound that is also used as an anti-microbial agent for meat products.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 1729-1729 null
Modeling the growth of Listeria monocytogenes in cured ready-to-eat processed meat products by manipulation of sodium chloride, sodium diacetate, potassium lactate, and product moisture content.
Seman DL, et al.
Journal of Food Protection, 65(4), 651-658 (2002)
Michael P Gotsbacher et al.
Chembiochem : a European journal of chemical biology, 21(10), 1433-1445 (2019-12-22)
This work aimed to undertake the in situ conversion of the terminal amine groups of bacterial desferrioxamine (DFO) siderophores, including desferrioxamine B (DFOB), to azide groups to enable downstream click chemistry. Initial studies trialed a precursor-directed biosynthesis (PDB) approach. Supplementing Streptomyces
Huiling Zhu et al.
Molecular nutrition & food research, 62(9), e1700814-e1700814 (2018-03-07)
Flaxseed oil is a rich source of α-linolenic acid (ALA), which is the precursor of the long-chain n-3 polyunsaturated fatty acids (PUFAs), including docosahexaenoic acid (DHA) and eicosapentaenoic acid (EPA). This study investigates the protective effect of flaxseed oil against
Albert N Ngo et al.
Pharmaceutical research, 33(2), 367-383 (2015-11-11)
It is hypothesized that sodium acetate (SA) can be used for in situ coating of drug loaded chitosan NPs for improved physico-chemical properties. Tenofovir (TFV) is used as a model drug. Uncoated chitosan NPs are prepared by ionic gelation. SA
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service