Skip to Content
Merck
All Photos(1)

Key Documents

W387401

Sigma-Aldrich

2-Methyl-1-propanethiol

≥95%, FG

Synonym(s):

Isobutyl mercaptan

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$310.00

$310.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
1 KG
$310.00

About This Item

Linear Formula:
(CH3)2CHCH2SH
CAS Number:
Molecular Weight:
90.19
FEMA Number:
3874
Beilstein:
1730890
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.173
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$310.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor density

3.1 (vs air)

vapor pressure

124 mmHg ( 37.8 °C)

Assay

≥95%

refractive index

n20/D 1.4385 (lit.)

bp

87-89 °C (lit.)

density

0.831 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

SMILES string

CC(C)CS

InChI

1S/C4H10S/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

BDFAOUQQXJIZDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Targeting the cerebrovascular large neutral amino acid transporter (LAT1) isoform using a novel disulfide-based brain drug delivery system.: This paper discusses a novel brain drug delivery system that targets the LAT1 transporter, using 2-Methyl-1-propanethiol in the design of disulfide-based prodrugs for enhanced delivery across the blood-brain barrier (Killian et al., 2007).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service