Skip to Content
Merck
All Photos(1)

Key Documents

W376507

Sigma-Aldrich

(−)-Myrtenyl acetate

≥95%

Synonym(s):

(1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-methyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O2
CAS Number:
Molecular Weight:
194.27
FEMA Number:
3765
Beilstein:
4309977
EC Number:
Council of Europe no.:
10887
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.302
NACRES:
NA.21
grade:
Fragrance grade
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Fragrance grade
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥95%

optical activity

[α]20/D −47°, neat

refractive index

n20/D 1.472 (lit.)

bp

134-135 °C/38 mmHg (lit.)

density

0.991 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

herbaceous; floral

storage temp.

2-8°C

SMILES string

CC(=O)OCC1=CCC2CC1C2(C)C

InChI

1S/C12H18O2/c1-8(13)14-7-9-4-5-10-6-11(9)12(10,2)3/h4,10-11H,5-7H2,1-3H3

InChI key

BKATZVAUANSCKN-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service