Skip to Content
Merck
All Photos(1)

Documents

W370304

Sigma-Aldrich

DL-3-Methyl-2-butanol

≥98%, FG

Synonym(s):

3-Methyl-2-butanol, Isopropyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(OH)CH3
CAS Number:
Molecular Weight:
88.15
FEMA Number:
3703
Beilstein:
1718800
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.111
NACRES:
NA.21

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

Assay

≥98%

refractive index

n20/D 1.409 (lit.)

bp

112 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity

SMILES string

CC(C)C(C)O

InChI

1S/C5H12O/c1-4(2)5(3)6/h4-6H,1-3H3

InChI key

MXLMTQWGSQIYOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascal Fuchsmann et al.
Journal of agricultural and food chemistry, 63(34), 7511-7521 (2015-08-01)
To establish the odor profiles of three differently fabricated commercial Swiss Tilsit cheeses, analyses were conducted using headspace solid-phase microextraction gas chromatography-mass spectrometry/pulsed flame photometric detection and gas chromatography-olfactometry to identify and quantitate volatile compounds. In addition, odor quality and
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service