Skip to Content
Merck
All Photos(1)

Key Documents

W348805

Sigma-Aldrich

Ethyl 2-methylpentanoate

≥98%, FG

Synonym(s):

Manzanate

Sign Into View Organizational & Contract Pricing

Select a Size

W348805-SAMPLE
$59.80
200 G
$83.70
4 KG
$1,440.00

$59.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W348805-SAMPLE
$59.80
200 G
$83.70
4 KG
$1,440.00

About This Item

Linear Formula:
CH3CH2CH2CH(CH3)CO2C2H5
CAS Number:
Molecular Weight:
144.21
FEMA Number:
3488
EC Number:
Council of Europe no.:
10616
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.526
NACRES:
NA.21
grade:
FG
Fragrance grade
biological source:
synthetic
food allergen:
no known allergens

$59.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

grade

FG
Fragrance grade

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥98%

refractive index

n20/D 1.403 (lit.)

bp

153-152 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; melon; fruity

SMILES string

CCCC(C)C(=O)OCC

InChI

1S/C8H16O2/c1-4-6-7(3)8(9)10-5-2/h7H,4-6H2,1-3H3

InChI key

HZPKNSYIDSNZKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeedeh Noushini et al.
Scientific reports, 10(1), 19799-19799 (2020-11-15)
Diverse methods have been used to sample insect semiochemicals. Sampling methods can differ in efficiency and affinity and this can introduce significant biases when interpreting biological patterns. We compare common methods used to sample tephritid fruit fly rectal gland volatiles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service