Skip to Content
Merck
All Photos(1)

Key Documents

W340618

Sigma-Aldrich

2-Isopropyl-5-methyl-2-hexenal

≥95%, stabilized, FG

Synonym(s):

5-methyl-2-propan-2-ylhex-2-enal, Isodihydrolavandulal

Sign Into View Organizational & Contract Pricing

Select a Size

W340618-SAMPLE-K
$54.60
100 G
$92.90
1 KG
$521.00

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W340618-SAMPLE-K
$54.60
100 G
$92.90
1 KG
$521.00

About This Item

Linear Formula:
(CH3)2CHCH2CH=C[CH(CH3)2]CHO
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3406
EC Number:
Council of Europe no.:
10361
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.107
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥95%

contains

α-tocopherol as stabilizer

refractive index

n20/D 1.452 (lit.)

bp

189 °C (lit.)

density

0.845 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; lavender; woody; herbaceous

SMILES string

[H]C(=O)C(=CCC(C)C)C(C)C

InChI

1S/C10H18O/c1-8(2)5-6-10(7-11)9(3)4/h6-9H,5H2,1-4H3

InChI key

IOLQAHFPDADCHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Isopropyl-5-methyl-2-hexenal is a volatile aroma constituent of malted barley[1] and roasted coffee beans.[2] It can be formed via Maillard reaction between leucine and 3-methylbutanal.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compounds contributing to the characteristic aroma of malted barley.
Beal AD & Mottram DS.
Journal of Agricultural and Food Chemistry, 42(12), 2880-2884 (1994)
Effects of pH, temperature, and reactant molar ratio on L-leucine and D-glucose Maillard browning reaction in an aqueous system.
Renn PT & Sathe SK
Journal of Agricultural and Food Chemistry, 45(10), 3782-3787 (1997)
Justyna Aszyk et al.
Journal of chromatography. A, 1547, 86-98 (2018-03-15)
Many flavour compounds that are present in e-liquids for e-cigarettes are responsible for specific tastes and smoking sensations for users. Data concerning content and specific types of flavours is often limited and unknown to users. The aim of the research
Steam volatile aroma constituents of roasted cocoa beans.
Van Praag M, et al.
Journal of Agricultural and Food Chemistry, 16(6), 1005-1008 (1968)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service