Skip to Content
Merck
All Photos(1)

Key Documents

W329810

Sigma-Aldrich

3-Mercapto-2-butanone solution

10 wt. % in triacetin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2SHCH3
CAS Number:
Molecular Weight:
104.17
FEMA Number:
3298
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.047

biological source

synthetic

concentration

10 wt. % in triacetin

refractive index

n20/D 1.4352

density

1.142 g/mL at 25 °C

application(s)

flavors and fragrances

Organoleptic

coffee

SMILES string

CC(S)C(C)=O

InChI

1S/C4H8OS/c1-3(5)4(2)6/h4,6H,1-2H3

InChI key

XLMPYCGSRHSSSX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service