Skip to Content
Merck
All Photos(1)

Key Documents

W327613

Sigma-Aldrich

Dipropyl trisulfide

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14S3
CAS Number:
Molecular Weight:
182.37
FEMA Number:
3276
Beilstein:
1736293
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Flavis number:
12.023
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

alliaceous; sulfurous

SMILES string

CCCSSSCCC

InChI

1S/C6H14S3/c1-3-5-7-9-8-6-4-2/h3-6H2,1-2H3

InChI key

GAZXPZNJTZIGBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service