Skip to Content
Merck
All Photos(1)

Key Documents

W322504

Sigma-Aldrich

Phenyl disulfide

≥98%

Synonym(s):

Diphenyl disulfide, NSC 2689

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SSC6H5
CAS Number:
Molecular Weight:
218.34
FEMA Number:
3225
Beilstein:
639794
EC Number:
Council of Europe no.:
11757
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.043

biological source

synthetic

grade

Halal
Kosher

Assay

≥98%

mp

58-60 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

earthy; sulfurous

SMILES string

S(Sc1ccccc1)c2ccccc2

InChI

1S/C12H10S2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H

InChI key

GUUVPOWQJOLRAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Mitchell et al.
Drug metabolism and drug interactions, 16(3), 191-206 (2000-12-16)
Radiolabelled [UL-14C]-diphenyl sulphide, [UL-14C]-diphenyl sulphoxide and [UL-14C]-diphenyl sulphone were administered by gavage (1.0 mmol/kg body weight) to adult male Wistar rats following an overnight fast. For all compounds, faeces were the major route of excretion of radioactivity (50%). Urinary elimination
Ana Mijovilovich et al.
The journal of physical chemistry. A, 114(35), 9523-9528 (2010-08-19)
Sulfur K-edge XANES was measured for two divalent sulfurs (dibenzyl and benzyl phenyl) and two disulfides (dibenzyl and diphenyl). The absorption spectra could be assigned using density functional theory with the "half core hole" approximation for the core hole including
C W Nogueira et al.
Toxicology, 191(2-3), 169-178 (2003-09-11)
Organochalcogens are important intermediates and useful reagents in organic synthesis, which can increase human exposure risk to these chemicals in the workplace. As well, there are a number of reported cases of acute toxicity following organochalcogen ingestion of vitamins and
J I Rossato et al.
Neurochemical research, 27(4), 297-303 (2002-04-18)
Ebselen (2-phenyl- 1,2-benzisoselenazole-3 (2H)-one) is a seleno-organic compound with antioxidant properties, and anti-inflammatory actions. Recently, ebselen improved the outcome of acute ischemic stroke in humans. In the present study, the potential antioxidant capacity of organochalcogenide compounds diphenyl diselenide (PhSe)2, diphenyl
Y Ito et al.
Mutation research, 393(3), 307-316 (1997-12-11)
The suppressive effect of S-methyl methanethiosulfonate (MMTS) on aflatoxin B1 (AFB1)- or methyl methanesulfonate (MMS)-induced chromosome aberrations (CA) in rat bone marrow cells was studied. MMTS significantly suppressed CA induced by both AFB1 (an indirect-acting carcinogen) and MMS (a direct-acting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service