Skip to Content
Merck
All Photos(2)

Key Documents

W280320

Sigma-Aldrich

3-Octanone

natural (US), ≥97%, FG

Synonym(s):

Ethyl amyl ketone, Ethyl pentyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COC2H5
CAS Number:
Molecular Weight:
128.21
FEMA Number:
2803
Beilstein:
1700021
EC Number:
MDL number:
UNSPSC Code:
12352200
Flavis number:
7.062
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.415 (lit.)

bp

167-168 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

butter; herbaceous; spicy

SMILES string

CCCCCC(=O)CC

InChI

1S/C8H16O/c1-3-5-6-7-8(9)4-2/h3-7H2,1-2H3

InChI key

RHLVCLIPMVJYKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service