W272418
Methyl nonanoate
≥97%
Synonym(s):
Methyl pelargonate, Nonanoic acid methyl ester
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
Kosher
Assay
≥97%
refractive index
n20/D 1.421 (lit.)
bp
213-214 °C (lit.)
density
0.875 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
waxy; fruity; sweet; wine-like
SMILES string
CCCCCCCCC(=O)OC
InChI
1S/C10H20O2/c1-3-4-5-6-7-8-9-10(11)12-2/h3-9H2,1-2H3
InChI key
IJXHLVMUNBOGRR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service