Skip to Content
Merck
All Photos(1)

Key Documents

W272000

Sigma-Aldrich

Methyl 3-(methylthio)propionate

≥98%, FG

Synonym(s):

Methyl 3-(methylmercapto)propionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2COOCH3
CAS Number:
Molecular Weight:
134.20
FEMA Number:
2720
Beilstein:
1745077
EC Number:
Council of Europe no.:
428
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.002
NACRES:
NA.21
Organoleptic:
garlic; onion; vegetable; sulfurous; tomato
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
(EU Regulation 1223/2009)
(follows IFRA guidelines)
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

(EU Regulation 1223/2009)
(follows IFRA guidelines)
meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.465 (lit.)

bp

74-75 °C/13 mmHg (lit.)

density

1.077 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

garlic; onion; vegetable; sulfurous; tomato

SMILES string

COC(=O)CCSC

InChI

1S/C5H10O2S/c1-7-5(6)3-4-8-2/h3-4H2,1-2H3

InChI key

DMMJVMYCBULSIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 3-(methylthio)propionate is a sulfur-containing flavor compound found in pineapple, muskmelon and strawberries.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The significance of volatile sulfur compounds in food flavors: An overview.
McGorrin RJ.
ACS Symp. Ser., 1068, 3-31 (2011)
High-impact sulfur-containing flavor compounds [J].
Bao-Guo S.
Food Sci. Technol., 7, 063-063 (2006)
Changes in strawberry volatile sulfur compounds due to genotype, fruit maturity and sample preparation.
Du X, et al.
Flavour and Fragrance Journal, 27(6), 398-404 (2012)
Volatile components in aqueous essence and fresh fruit of Cucumis melo cv. Athena (muskmelon) by GC-MS and GC-O.
Jordan MJ, et al.
Journal of Agricultural and Food Chemistry, 49(12), 5929-5933 (2001)
Yuwadee Ackarabanpojoue et al.
Journal of food science, 80(5), E998-1004 (2015-04-02)
This study aimed at investigating the effect of nitrate removal from pineapple juice by electrodialysis (ED) on selected properties of the ED-treated juice. Single-strength pineapple juice with reduced pulp content was treated by ED to reduce the nitrate concentration to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service