Skip to Content
Merck
All Photos(1)

Documents

W266906

Sigma-Aldrich

Menthyl isovalerate

≥98%, FG

Synonym(s):

Menthyl 3-methylbutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
FEMA Number:
2669
EC Number:
Council of Europe no.:
450
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.455
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

optical activity

[α]20/D −64°, neat

bp

260-262 °C/750 mmHg (lit.)

density

0.909 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; cooling; woody; sweet

SMILES string

CC(C)CC(=O)OC1CC(C)CCC1C(C)C

InChI

1S/C15H28O2/c1-10(2)8-15(16)17-14-9-12(5)6-7-13(14)11(3)4/h10-14H,6-9H2,1-5H3

InChI key

VYQSSWZYPCCBRN-UHFFFAOYSA-N

General description

Menthyl isovalerate is one of the key constituents of peppermint oil. It is also reported in the essential oils of some Tanacetum species.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I P Bondarenko et al.
Likars'ka sprava, (3-4)(3-4), 110-113 (1996-03-01)
As many as 20 patients with neurocirculatory dystonia (NCD) and 10 IHD patients presenting with stable exertional angina were evaluated for an effectiveness of antianginal action of validol tablets commercially- and noncommercially produced, the above tablets being of the changed
Essential oils of tanacetum elburensis mozaff. And tanacetum persicum (boiss.) mozaff. From Iran.
Habibi Z, et al.
J. Essent. Oil Res., 19(4), 310-312 (2007)
Medicinally potential plants of Labiatae (Lamiaceae) family: an overview.
Raja RR.
Research journal of medicinal plant, 6(3), 203-213 (2012)
H Leclerc et al.
Journal of dairy science, 91(8), 3179-3183 (2008-07-25)
The validity of national genetic evaluations depends on the quality of input data, on the model of analysis, and on the correctness of genetic evaluation software. A general strategy was developed to validate national breeding value prediction software: performances from
A M López-Parra et al.
Clinical biochemistry, 46(1-2), 167-169 (2012-08-25)
We have developed a genotyping system to determine the alleles of genes related to interindividual variability in acenocoumarol dosage requirements. This genotyping system is intended for routine clinical use and therefore it is essential that it be simple, fast and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service