Skip to Content
Merck
All Photos(1)

Key Documents

W252204

Sigma-Aldrich

Ginger oil

natural, FCC, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W252204-SAMPLE-K
$62.70
250 G
$147.00
1 KG
$322.00

$62.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W252204-SAMPLE-K
$62.70
250 G
$147.00
1 KG
$322.00

About This Item

CAS Number:
FEMA Number:
2522
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
biological source:
Zingiber officinale Roscoe
food allergen:
no known allergens

$62.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

Zingiber officinale Roscoe

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D −39°, neat

refractive index

n20/D 1.49 (lit.)

bp

254 °C (lit.)

density

0.871 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

ginger; citrus; woody; spicy

Related Categories

General description

Ginger oil generally contains sesquiterpene hydrocarbons such as zingiberene, ar-curcumene, β-bisabolene and β-sesquiphellandrene and monoterpenes such as geranial, neral and camphene.[1]

Biochem/physiol Actions

Taste at 30 ppm

Preparation Note

Extraction method: steam distillation

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Essential oil composition of diploid and tetraploid clones of ginger (Zingiber officinale Roscoe) grown in Australia.
Wohlmuth H, et al.
Journal of Agricultural and Food Chemistry, 54(4), 1414-1419 (2006)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 66-66 (2005)
Mosciano, G.
Perfumer & Flavorist, 24, 50-50 (1999)

Articles

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service