Skip to Content
Merck
All Photos(1)

Documents

W244740

Sigma-Aldrich

Ethyl nonanoate

natural, 98%, FG

Synonym(s):

Ethyl pelargonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COOC2H5
CAS Number:
Molecular Weight:
186.29
FEMA Number:
2447
Beilstein:
1759169
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.107
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

vapor pressure

0.08 mmHg ( 25 °C)

Assay

98%

refractive index

n20/D 1.422 (lit.)

bp

119 °C/23 mmHg (lit.)

density

0.866 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

fragrance allergen

no known allergens

Organoleptic

oily; nutty; fruity

SMILES string

CCCCCCCCC(=O)OCC

InChI

1S/C11H22O2/c1-3-5-6-7-8-9-10-11(12)13-4-2/h3-10H2,1-2H3

InChI key

BYEVBITUADOIGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service