Skip to Content
Merck
All Photos(3)

Key Documents

W225299

Sigma-Aldrich

β-Caryophyllene

greener alternative

natural, ≥80%, FG

Synonym(s):

(−)-trans-Caryophyllene, trans-(1R,9S)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0]undec-4-ene

Sign Into View Organizational & Contract Pricing

Select a Size

W225299-SAMPLE-K
$56.00
1 KG
$117.00
9 KG
$526.00

$56.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W225299-SAMPLE-K
$56.00
1 KG
$117.00
9 KG
$526.00

About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
FEMA Number:
2252
Beilstein:
2044564
MDL number:
UNSPSC Code:
12164502
Flavis number:
1.007
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
food allergen:
no known allergens

$56.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

botanical

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥80%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.5 (lit.)

bp

129-130 °C/14 mmHg (lit.)

density

0.901 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

β-caryophyllene (ox.)

greener alternative category

Organoleptic

woody; spicy

SMILES string

C\C1=C/CCC(=C)[C@H]2CC(C)(C)[C@@H]2CC1

InChI

1S/C15H24/c1-11-6-5-7-12(2)13-10-15(3,4)14(13)9-8-11/h6,13-14H,2,5,7-10H2,1,3-4H3/b11-6+/t13-,14-/m1/s1

InChI key

NPNUFJAVOOONJE-GFUGXAQUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How large is sample K size?

    1 answer
    1. The SAMPLE-K quantities vary from product to product and lot to lot. There is no established quantity assigned. These are typically gram quantities intended for use in formulation evaluation, however, some more expensive compounds may contain smaller amounts.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service