Skip to Content
Merck
All Photos(1)

Key Documents

W213209

Sigma-Aldrich

Benzoin

≥98%

Synonym(s):

α-Hydroxy-α-phenylacetophenone, α-Hydroxybenzyl phenyl ketone, 2-Hydroxy-2-phenylacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)COC6H5
CAS Number:
Molecular Weight:
212.24
FEMA Number:
2132
Beilstein:
391839
EC Number:
Council of Europe no.:
162
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.028

biological source

synthetic

grade

Kosher

reg. compliance

FDA 21 CFR 172.515
FDA 21 CFR 73.1

Assay

≥98%

bp

194 °C/12 mmHg (lit.)

mp

134-138 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

medicinal; vanilla; balsamic

SMILES string

OC(c1ccccc1)C(=O)c2ccccc2

InChI

1S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15H

InChI key

ISAOCJYIOMOJEB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarvesh Kumar et al.
Archiv der Pharmazie, 345(5), 368-377 (2011-12-23)
The interaction between leukocytes and the vascular endothelial cells (EC) via cellular adhesion molecules plays an important role in the pathogenesis of various inflammatory and autoimmune diseases. Small molecules that block these interactions have been targeted as potential therapeutic agents
Direct copper-catalyzed α-arylation of benzyl phenyl ketones with aryl iodides: route towards tamoxifen.
Grégory Danoun et al.
Angewandte Chemie (International ed. in English), 51(51), 12815-12819 (2012-11-13)
Jun He et al.
Electrophoresis, 32(10), 1164-1175 (2011-04-19)
In the present work we report, for the first time, the successful on-line coupling of chiral MEKC (CMEKC) to atmospheric pressure photoionization MS (APPI-MS). Four structurally similar neutral test solutes (e.g. benzoin (BNZ) derivatives) were successfully ionized by APPI-MS. The
Yankai Liu et al.
Organic letters, 14(5), 1310-1313 (2012-03-03)
Expanding upon the recently developed aminocatalytic asymmetric indole-2,3-quinodimethane strategy, a straightforward synthesis of structurally and stereochemically complex tetrahydrocarbazoles has been devised. The chemistry's complexity-generating power was further harnessed by designing a multicatalytic, one-pot Diels-Alder/benzoin reaction sequence to stereoselectively access trans-fused
Total synthesis and absolute stereochemistry of seragakinone A.
Akiomi Takada et al.
Angewandte Chemie (International ed. in English), 50(10), 2297-2301 (2011-02-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service