Skip to Content
Merck
All Photos(3)

Documents

W211233

Sigma-Aldrich

Melissa oil

greener alternative

natural, FG, product of Essential oils and Herbs Ltd

Synonym(s):

Balm leaves oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2112
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Melissa officinalis

Quality Level

grade

FG
Fragrance grade
natural

Agency

EU Regulation 1223/2009
follows IFRA guidelines

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

composition

Contains IFRA restricted Balm leaves extract, EU 1223/2009 restrictedLimonene and linalool, and EU 1223/2009 and IFRA restricted Citral,Citronellol, Geraniol, Eugenol, and Benzyl Benzoate.

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

citral (neral+geranial), linalool, citronellol, limonene (sum of D, L and DL), eugenol, benzyl benzoate, limonene (sum of D, L and DL), geraniol

greener alternative category

Organoleptic

fresh; citrus; sweet

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F

Flash Point(C)

80 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Essential oils are complex natural extracts consisting of aromatic compounds and related organic chemicals. While they hold great value as fragrance and flavor ingredients, their significant risk of adulteration necessitates consistent formulator testing for quality, naturalness, and allergen levels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service