Skip to Content
Merck
All Photos(1)

Documents

W207403

Sigma-Aldrich

Amyl hexanoate

≥98%, FG

Synonym(s):

n-Pentyl hexanoate, Amyl caproate, Hexanoic acid pentyl ester, NSC 46119, NSC 53794, Pentyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2(CH2)4CH3
CAS Number:
Molecular Weight:
186.29
FEMA Number:
2074
EC Number:
Council of Europe no.:
315
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.065
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.42 (lit.)

bp

226 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; waxy; sweet

SMILES string

CCCCCOC(=O)CCCCC

InChI

1S/C11H22O2/c1-3-5-7-9-11(12)13-10-8-6-4-2/h3-10H2,1-2H3

InChI key

WRFZKAGPPQGDDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Amyl hexanoate is a volatile flavor compound that is reported to occur in mayhaw fruit, apricot, and banana.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile constituents of apricot (Prunus armeniaca).
Takeoka GR, et al.
Journal of Agricultural and Food Chemistry, 38(2), 471-477 (1990)
Identification of volatile compounds from ripe mayhaw fruit (Crataegus opaca, C. aestivalis, and C. rufula).
Horvat RJ, et al.
Journal of Food Quality, 14(4), 307-312 (1991)
Biogenesis of banana volatiles.
Tressl R & Drawert F.
Journal of Agricultural and Food Chemistry, 21(4), 560-565 (1973)
Wei Wu et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(12), 1855-1863 (2015-09-12)
The multispecific organic anion drug transporters OAT6 (SLC22A20) and OAT1 (SLC22A6) are expressed in nasal epithelial cells and both can bind odorants. Sequence analysis of OAT6 revealed an evolutionarily conserved 79-amino acid (AA) fragment present not only in OAT6 but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service