Key Documents
TEM-Q250AR1.3
Sigma-Aldrich
QUANTIFOIL R 1.2/1.3
Gold Grids, size 200 mesh, pack of 50
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-Q250AR1.3 is currently not available for purchase. Contact Technical Service
packaging
pack of 50
size
200 mesh
General description
QUANTIFOIL with circular holes is used in cryoelectron tomographic reconstruction. The roundness of the holesis advantageous with respect to the formation of an icelayer of constant thickness. Assessment of the image quality is easier when foil is incl
Choose from one of the most recent versions:
Certificates of Analysis (COA)