Key Documents
TEM-G1500HHCU
Sigma-Aldrich
1500 Mesh Hexagonal Copper Grid
vial of 15
Synonym(s):
HEXAGONAL MESH 1500#, 15/VIAL
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-G1500HHCU is currently not available for purchase. Contact Technical Service
packaging
vial of 15
General description
High Mesh Value Standard Hexagonal Mesh Grids: Copper Grid, 1500 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 16.5 μm, hole ~10.5μm, bar ~6μm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)