Key Documents
TEM-FF100CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 100 mesh, box of 50
Synonym(s):
FORMVAR SQUARE MESH - CU 100 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-FF100CUUL is currently not available for purchase. Contact Technical Service
packaging
box of 50
size
100 mesh
General description
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 100 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)