Key Documents
TEM-CF100HCU
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 100 mesh, box of 25
Synonym(s):
CARBON ONLY ON HEX COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-CF100HCU is currently not available for purchase. Contact Technical Service
material
carbon film
copper grid
description
film layer thickness 5 - 6 nm carbon
packaging
box of 25
size
100 mesh
General description
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 100 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)