Skip to Content
Merck
All Photos(1)

Key Documents

T75752

Sigma-Aldrich

3,3,5-Trimethylcyclohexanone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$50.30
1 L
$153.00

$50.30

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$50.30
1 L
$153.00

About This Item

Linear Formula:
(CH3)3C6H7(=O)
CAS Number:
Molecular Weight:
140.22
Beilstein:
507452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$50.30

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

98%

refractive index

n20/D 1.445 (lit.)

bp

188-192 °C (lit.)

mp

−10 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

SMILES string

CC1CC(=O)CC(C)(C)C1

InChI

1S/C9H16O/c1-7-4-8(10)6-9(2,3)5-7/h7H,4-6H2,1-3H3

InChI key

POSWICCRDBKBMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F

Flash Point(C)

64 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Af Delgove et al.
Journal of chemical technology and biotechnology (Oxford, Oxfordshire : 1986), 93(8), 2131-2140 (2018-08-03)
It is widely accepted that the poor thermostability of Baeyer-Villiger monooxygenases limits their use as biocatalysts for applied biocatalysis in industrial applications. The goal of this study was to investigate the biocatalytic oxidation of 3,3,5-trimethylcyclohexanone using a thermostable cyclohexanone monooxygenase
Marie A F Delgove et al.
Organic process research & development, 22(7), 803-812 (2018-10-03)
Although Baeyer-Villiger monooxygenases (BVMOs) have gained attention in recent years, there are few cases of their upscaled application for lactone synthesis. A thermostable cyclohexanone monooxygenase from Thermocrispum municipale (TmCHMO) was applied to the oxidation of 3,3,5-trimethylcyclohexanone using a glucose dehydrogenase
Coraline Simon et al.
Toxicology in vitro : an international journal published in association with BIBRA, 37, 121-133 (2016-10-19)
Endocrine activity of 65 compounds migrating from polycarbonate replacement plastic baby bottles was assessed using in vitro cell based assays (reporter gene assays) involving 7 nuclear receptors, i.e. human steroid hormones receptors (oestrogen, androgen, progesterone and glucocorticoid receptors), human thyroid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service