Skip to Content
Merck
All Photos(1)

Key Documents

T54704

1,1,1-Trichloroethane

97%

Synonym(s):

Methylchloroform, ‘Chlorothene’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCH3
CAS Number:
Molecular Weight:
133.40
Beilstein:
1731614
MDL number:
PubChem Substance ID:

vapor density

4.6 (vs air)

vapor pressure

100 mmHg ( 20 °C)

Assay

97%

autoignition temp.

998 °F

contains

1-3% 1,4-dioxane as stabilizer

expl. lim.

7.5-15 %, 25 °F

refractive index

n20/D 1.4366 (lit.)

bp

74-76 °C (lit.)

mp

−35 °C (lit.)

density

1.336 g/mL at 20 °C (lit.)

SMILES string

CC(Cl)(Cl)Cl

InChI

1S/C2H3Cl3/c1-2(3,4)5/h1H3

InChI key

UOCLXMDMGBRAIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Warning

1,4-Dioxane is a cancer suspect agent

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Ozone 1

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew R Babbin et al.
Nature communications, 11(1), 3672-3672 (2020-07-30)
Nitrous oxide (N2O) is a powerful greenhouse gas and ozone depleting substance, but its natural sources, especially marine emissions, are poorly constrained. Localized high concentrations have been observed in the oxygen minimum zones (OMZs) of the tropical Pacific but the
Sigit Purbadi et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 1986793-1986793 (2020-06-23)
Cervical cancer is a leading cause of death among women worldwide, particularly in Indonesia. The main treatment of advanced-stage cervical cancer is radiation; however, the outcomes do not meet the required expectations. [1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5dione] has been reported in several studies for
Mario Vincenzo Russo et al.
International journal of environmental research and public health, 18(4) (2021-02-12)
This paper shows a characterization of the organic and inorganic fraction of river waters (Tiber and Marta) and ores/soil samples collected in the Northern Latium region of Italy for evaluating the anthropogenic/natural source contribution to the environmental pollution of this
Sara Kleindienst et al.
Systematic and applied microbiology, 40(3), 150-159 (2017-03-16)
Taxonomic assignments of anaerobic dichloromethane (DCM)-degrading bacteria remain poorly constrained but are important for understanding the microbial diversity of organisms contributing to DCM turnover in environmental systems. We describe the taxonomic classification of a novel DCM degrader in consortium RM
Hang Yin et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(14), 1903381-1903381 (2020-07-28)
The development of gene delivery has attracted increasing attention, especially when the introduction and application of the CRISPR/Cas9 gene editing system appears promising for gene therapy. However, ensuring biosafety and high gene editing efficiency at the same time poses a great challenge for its in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service